RCM with Na Lactate, Hardy Diagnostics
Catalog # 89497-622
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Tube
- Description:RCM with Na Lactate
- Size:16x125mm
- Storage Temperature:2-8C
- Minimum Order Quantity:5
- Cat. No.:89497-622
- Application:For the cultivation and enumeration of clostridia, and other anaerobic and facultative bacteria from foods and clinical specimens.
- Supplier no.:K351
Specifications
About this item
RCM with Na Lactate (Reinforced Clostridial Medium with Sodium Lactate), 16x125mm tube, order by the package of 20 tubes, by Hardy Diagnostics. MINIMUM ORDER: 5 packs.
For the cultivation and enumeration of clostridia, and other anaerobic and facultative bacteria from foods and clinical specimens.
RCM with Na Lactate (Reinforced Clostridial Medium with Sodium Lactate), 16x125mm tube, order by the package of 20 tubes, by Hardy Diagnostics
MINIMUM ORDER: 5 packs.