Fluid Thioglycollate, Hardy Diagnostics
Catalog # 89497-614
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Tube
- Size:16x125mm
- Storage Temperature:2-30C
- Cat. No.:89497-614
- Fill volume:10 ml
- Supplier no.:K282
Specifications
About this item
Fluid Thioglycollate, USP, 16x125mm glass tube with needle-port hungate cap, 10ml fill, order by the package of 20, by Hardy Diagnostics.
- For use in the cultivation of aerobic, microaerophilic, and anaerobic microorganisms in normally sterile materials.
- Contains a resazurin oxygen indicator. Has a rubber septum in the screw cap for inoculation by needle.
- Not intended for human diagnostic use. For sterility and environmental testing only.
Certifications: USP
Ordering information: Not intended for human diagnostic use. See catalog number 89407-974 for a Fluid Thio for diagnostic use (medical device).