Order Entry
Puerto Rico
ContactUsLinkComponent
Eisco® Enlarged Human Larynx Model, 3 Parts
Eisco® Enlarged Human Larynx Model, 3 Parts
Catalog # 470212-462
Supplier:  Wards
undefined
Eisco® Enlarged Human Larynx Model, 3 Parts
Catalog # 470212-462
Supplier:  Wards
Supplier Number:  AM270AS

Specifications

  • Description:
    Larynx
  • Height:
    26 cm
  • Length:
    13 cm
  • Cat. No.:
    470212-462
  • Width:
    13 cm
  • Supplier no.:
    AM270AS
  • No. of parts:
    3

Specifications

About this item

Enlarged model of the human larynx, including voice box.

  • Enlarged (3x) model of human larynx
  • 3 parts, including voice box
  • Hand painted
  • Mounted on base
  • Includes numbered key card

The three part model human larynx allows students to dissect the larynx and observe the movements of the epiglottis and cartilage in the voice box.