ContactUsLinkComponent
Eisco® Electric Tuning Fork
Eisco® Electric Tuning Fork
Catalog # 470138-868
Supplier:  Wards
undefined
Eisco® Electric Tuning Fork
Catalog # 470138-868
Supplier:  Wards
Supplier Number:  PH0746CNC
Specifications
  • Description:
    Electric Tuning Fork
  • Cat. No.:
    470138-868
  • Supplier no.:
    PH0746-CNC
Specifications
About this item
Show relationships between frequency, length, tension and mass
  • Heavy Duty
  • Repeatability
Learn about Melde's standing wave experiments with this fork. An electromagnet drives the fork to a sustained frequency of approx. 80Hz. One prong of the tuning fork has a captive knurled thumbscrew for performing Melde's experiment. Amplitude of the tuning fork vibrations is varied by means of a threaded adjusted screw. Universal binding posts are finished to provide electrical power from an external source of 2 to 6 volts DC, required but not included.