ContactUsLinkComponent
Modified Letheen Broth, BD Biosciences
Modified Letheen Broth, BD Biosciences
Catalog # 90003-730
Supplier:  BD
CAS Number:  
Modified Letheen Broth, BD Biosciences
Catalog # 90003-730
Supplier:  BD
Supplier Number:  263010
CAS Number:  
Specifications
  • Cat. No.:
    90003-730
  • Supplier no.:
    263010
Specifications
About this item
Dehydrated Culture Media for microbiological evaluation of cosmetics
Inactivates preservative agents.