ContactUsLinkComponent
Starfish Universal 5-Way Clamp
Starfish Universal 5-Way Clamp
Catalog # 47800-064
Supplier:  Heidolph NA, LLC
Starfish Universal 5-Way Clamp
Catalog # 47800-064
Supplier:  Heidolph NA, LLC
Supplier Number:  015890310
Specifications
  • Description:
    Starfish universal 5-way clamp with velcro fasteners
  • Cat. No.:
    47800-064
  • Supplier no.:
    15890310
Specifications
About this item
The StarFish clamp allows you to hold glassware of virtually any size and comes with Velcro support straps.
Each clamp features 5 telescopic arms that can be extended and locked in place to suit your needs.