ContactUsLinkComponent
Microcentrifuge Storage Box, Simport Scientific
Microcentrifuge Storage Box, Simport Scientific
  89499-338
Microcentrifuge Storage Box, Simport Scientific
  89499-338
 :  T330-64
  • Color:
    Blue
  • Description:
    Microcentrifuge Storage Box
  • Cat. No.:
    89499-338
  • Supplier no.:
    T330-64
This Microcentrifuge Storage Box with a polyurethane foam insert is ideal for holding up to 64 tubes from 0.5 mL to 2 mL
Can withstand temperature range of -90 to 80°C. A transparent cover allows you to see the contents of the box, and is keyed to the base in order to prevent misalignment. Stackable. Made of high impact polystyrene.