Order Entry
Puerto Rico
ContactUsLinkComponent
 
Phenazine Paper, DLI, Decon Labs
  89423-136
 :  
undefined
Phenazine Paper, DLI, Decon Labs
  89423-136
 :  1201
 :  

 

  • Description:
    5 yards per roll, 0.25" wide
  • Cat. No.:
    89423-136
  • Range:
    pH 4.5 - 7.5
  • Supplier no.:
    1201

 

 

Decon's phenazine paper is specifically designed for determining pH in the range of 4.0 to 8.0.

  • Color scale chart is graduated in 0.5 pH units

Each phenazine paper roll includes over 16 feet of paper spooled on plastic dispenser. This easily read paper gives sharp, clear color changes for easy comparison to the color scale provided on the dispenser.