Vacuum Trap with #15 O-Ring Joints, Ace Glass Incorporated
89428-016
Product available on GSA Advantage®
Small Business Enterprise
- Description:Vacuum Trap Flange Connection
- Cat. No.:89428-016
- Supplier no.:8771-100
This borosilicate glass vacuum trap is supplied with a no
15 O-Ring side connecting joint. Ideal for vacuum cart and other vacuum or NO-AIR applications. Supplied complete with stainless steel ball-joint clamp and FETFE O-Ring.