Order Entry
Puerto Rico
ContactUsLinkComponent
Vanillin for synthesis, Sigma-Aldrich®
Vanillin for synthesis, Sigma-Aldrich®
Catalog # EM8.18718.0500
Supplier:  MilliporeSigma
CAS Number:  
undefined
Vanillin for synthesis, Sigma-Aldrich®
Catalog # EM8.18718.0500
Supplier:  MilliporeSigma
Supplier Number:  8.18718.0500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g
  • Cat. No.:
    EM8.18718.0500
  • Supplier no.:
    8.18718.0500

Specifications

Specification Test Results

  • Solubility:
    10 g/l (25 °C)
  • Melting Point:
    82 °C
  • Molar Mass:
    152.14 g/mol
  • Bulk Density:
    600 kg/m3
  • Boiling Point:
    154 °C (13 hPa)
  • Vapor Pressure:
    0.0029 hPa (25 °C)
  • Flash Point:
    153 °C
  • Density:
    1.06 g/cm3 (20 °C)
  • pH:
    4.3 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    >400 °C

Specification Test Results

About this item

 :
 :152.15 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:121-33-5
Formula:C₈H₈O₃
MDL Number:MFCD00006942