Order Entry
Puerto Rico
ContactUsLinkComponent
Methyl acetate for synthesis, Sigma-Aldrich®
Methyl acetate for synthesis, Sigma-Aldrich®
Catalog # EM8.09711.2500
Supplier:  MilliporeSigma
CAS Number:  
Methyl acetate for synthesis, Sigma-Aldrich®
Catalog # EM8.09711.2500
Supplier:  MilliporeSigma
Supplier Number:  8.09711.2500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. No.:
    EM8.09711.2500
  • Supplier no.:
    8.09711.2500

Specifications

Specification Test Results

  • Solubility:
    319 g/l (20 °C)
  • Melting Point:
    -98 °C
  • Molar Mass:
    74.08 g/mol
  • Boiling Point:
    56 - 58 °C (1013 hPa)
  • Vapor Pressure:
    217 hPa (20 °C)
  • Flash Point:
    -13 °C
  • Refractive Index:
    1.3614 (20 °C, 589 nm)
  • Explosion Limit:
    3.1 - 16 %(V)
  • Density:
    0.93 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    455 °C
  • Saturation concentration:
    665 g/m3 (20 °C) Air
  • Evaporation number:
    2.2
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

About this item

 :
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :74.08 g/mol
UN:1231
CAS Number:79-20-9
Formula:C₃H₆O₂
MDL Number:MFCD00008711