Order Entry
Puerto Rico
ContactUsLinkComponent
Tetraethylene glycol for synthesis, Sigma-Aldrich®
Tetraethylene glycol for synthesis, Sigma-Aldrich®
Catalog # EM8.08619.1000
Supplier:  MilliporeSigma
CAS Number:  
Tetraethylene glycol for synthesis, Sigma-Aldrich®
Catalog # EM8.08619.1000
Supplier:  MilliporeSigma
Supplier Number:  8.08619.1000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 L
  • Cat. No.:
    EM8.08619.1000
  • Supplier no.:
    8.08619.1000

Specifications

Specification Test Results

  • Solubility:
    1000 g/l (20 °C)
  • Melting Point:
    -5 °C
  • Molar Mass:
    194.22 g/mol
  • Boiling Point:
    327 - 328 °C (1013 hPa)
  • Vapor Pressure:
    0.01 hPa (20 °C)
  • Flash Point:
    174 °C
  • Refractive Index:
    1.4609 (20 °C, 589 nm)
  • Explosion Limit:
    0.5 - 3.4 %(V)
  • Density:
    1.12 g/cm3 (20 °C)
  • pH:
    8.5 - 9.0 (500 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :194.23 g/mol
 :M-Clarity™ quality level = MQ200
CAS Number:112-60-7
Formula:C₈H₁₈O₅
MDL Number:MFCD00002879