Lithium metaborate, Spectromelt® A20 for XRF (X-ray fluorescence), fused, Supelco®
Catalog # EM1.12996.1000
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:EM1.12996.1000
- Supplier no.:1.12996.1000
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:840 °C
- Molar Mass:49.75 g/mol
- Bulk Density:1300 kg/m3
Specification Test Results
About this item
:
:49.75 g/mol
Merck Index:13,05546
Boiling Pt:1719 °C (1013 hPa)
Melting Pt:845 °C
Density:1.4 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:236-631-5
CAS Number:13453-69-5
Formula:BLiO₂
MDL Number:MFCD00011089
Boiling Pt:1719 °C (1013 hPa)