


Yttrium (III) nitrate hexahydrate, Sigma-Aldrich®
Catalog # EM1.12516.0025
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:25 g
- Cat. No.:EM1.12516.0025
- Supplier no.:1.12516.0025
Specifications
Specification Test Results
- Solubility:>1500 g/l (20 °C)
- Molar Mass:383.01 g/mol
- Bulk Density:1100 kg/m3
- Density:1.1 g/cm3 (20 °C)
- pH:3.5 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:383.01 g/mol
Melting Pt:51.8 °C
Density:2.682 g/cm³ (20 °C)
ADR:5.1,
:M-Clarity™ quality level = MQ200
EINECS:233-802-6
UN:1477
:GHS03
:GHS05
:GHS07
CAS Number:13494-98-9
Formula:N₃O₉Y·6H₂O
MDL Number:MFCD00149940