
Hydrochloric acid 0.1 M concentrated aqueous solution, Titrisol®, Supelco®
Catalog # EM1.09973.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Cardboard box
- Size:1 piece
- Cat. No.:EM1.09973.0001
- Supplier no.:1.09973.0001
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.03 g/cm3 (20 °C)
- pH:< - 1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,
:36.46
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324