
Sodium hydroxide 0.25 M concentrated aqueous solution, Titrisol®, Supelco®
Catalog # EM1.09958.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Cardboard box
- Size:1 pack
- Cat. No.:EM1.09958.0001
- Supplier no.:1.09958.0001
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.185 g/cm3 (20 °C)
- pH:14 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1.327 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548