Silicon Standard, Titrisol®, MilliporeSigma
Catalog # EM1.09947.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:14% NaOH
- Pack type:Plastic Ampule
- Size:1 Ampule
- Cat. No.:EM1.09947.0001
- Element:Silicon
- Supplier no.:1.09947.0001