

Potassium hydroxide 1 N in aqueous solution, Titrisol®, Supelco®
Catalog # EM1.09918.0001
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Cardboard box
- Size:1 pack
- Cat. No.:EM1.09918.0001
- Supplier no.:1.09918.0001
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.58 g/cm3 (20 °C)
- pH:>13.5 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,
:GHS05
:GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553