MColortest™ pH-indicator solution pH 4.0 - 10.0 Universal Indicator with colour Card
Supplier: MilliporeSigma
Certificates
About this item
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
- Results in approx. 1 Minute
- Easy to use
- Application-oriented measuring ranges
- Portable
Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Related Information
Certificates
Search by Lot Numberghs_1.09175
SDS
MilliporeSigma Brewery
Product Literature
VWR GHS Compliance Letter (US)