1-(2-Pyridylazo)-2-naphthol (PAN) indicator for metal titration, Supelco®
Catalog # EM1.07531.0005
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:5 g
- Cat. No.:EM1.07531.0005
- Supplier no.:1.07531.0005
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:136 - 141 °C
- Molar Mass:249.27 g/mol
- Bulk Density:190 kg/m3
Specification Test Results
About this item
:
:249.27 g/mol
Boiling Pt:412 °C (1013 hPa)
Melting Pt:137…140 °C
Storage Temperature:Ambient
EINECS:201-637-9
CAS Number:85-85-8
Formula:C₁₅H₁₁N₃O
MDL Number:MFCD00004071