Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium tetraoxalate dihydrate, Certipur® certified sec. standard reference buffer solution, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®
Irritant
Potassium tetraoxalate dihydrate, Certipur® certified sec. standard reference buffer solution, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®
Catalog # EM1.07204.0105
Supplier:  MilliporeSigma
CAS Number:  
Irritant
Potassium tetraoxalate dihydrate, Certipur® certified sec. standard reference buffer solution, directly traceable to primary reference material (SRM) from NIST/PTB, Supelco®
Catalog # EM1.07204.0105
Supplier:  MilliporeSigma
Supplier Number:  1.07204.0105
CAS Number:  

Specifications

  • Pack type:
    Cardboard box
  • Size:
    1 pack
  • Cat. No.:
    EM1.07204.0105
  • Supplier no.:
    1.07204.0105

Specifications

Specification Test Results

  • Density:
    1.003 g/cm3 (20 °C)
  • pH:
    1.7 (H2O, 25 °C)

Specification Test Results

About this item

 :
 :254.19 g/mol (218.16 anhy)
Storage Temperature:Ambient
EINECS:204-874-6
UN:3288
ADR:6.1,III
 :GHS07
CAS Number:6100-20-5
Formula:C₄H₃KO₈·2H₂O
MDL Number:MFCD00150443