Order Entry
Puerto Rico
ContactUsLinkComponent
Ninhydrin solution in 2-propanol, spray reagent for TLC, Supelco®
Flammable
Irritant
Ninhydrin solution in 2-propanol, spray reagent for TLC, Supelco®
Catalog # EM1.06705.0100
Supplier:  MilliporeSigma
CAS Number:  
undefined
Flammable
Irritant
Ninhydrin solution in 2-propanol, spray reagent for TLC, Supelco®
Catalog # EM1.06705.0100
Supplier:  MilliporeSigma
Supplier Number:  1.06705.0100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    EM1.06705.0100
  • Supplier no.:
    1.06705.0100

Specifications

About this item

 :
 :178.14 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ100
UN:1219
 :GHS02
 :GHS07
CAS Number:485-47-2
Formula:C₉H₆O₄
MDL Number:MFCD00003791