Sodium chloride ≥99.5% (silver nitrate titration) [99-100.5% dry basis (silver nitrate titration)], EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1461 °C (1013 hPa)
Melting Pt:801 °C (lit.)
Density:2.16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Specification Test Results
- Solubility:358 g/l (20 °C)
- Melting Point:801 °C
- Molar Mass:58.44 g/mol
- Bulk Density:1140 kg/m3
- Boiling Point:1461 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (865 °C)
- Density:2.17 g/cm3 (20 °C)
- pH:6 - 9 (50 g/l, H2O, 20 °C)