Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
Lithium sulfate monohydrate, EMSURE® for analysis, Supelco®
  EM1.05694.0250
 :  
 
Lithium sulfate monohydrate, EMSURE® for analysis, Supelco®
  EM1.05694.0250
 :  1.05694.0250
 :  

 

  • Pack type:
    Plastic bottle
  • Size:
    250 g
  • Cat. No.:
    EM1.05694.0250
  • Supplier no.:
    1.05694.0250

 

 

  • Solubility:
    340 g/l (20 °C)
  • Melting Point:
    120 °C
  • Molar Mass:
    127.96 g/mol
  • Bulk Density:
    940 kg/m3
  • Density:
    2.05 g/cm3 (20 °C)
  • pH:
    4.5 - 6.0 (50 g/l, H2O, 20 °C)

 

 

 :
 :127.96 g/mol
 :120 °C
 :2.05 g/cm³ (20 °C)
 :233-820-4
 :GHS07
 :Ambient
 :10102-25-7
 :Li₂SO₄·H₂O
 :MFCD00149766