
Sodium hydroxide 21% in aqueous solution analytical reagent, Supelco®
Catalog # EM1.05593.9025
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic container
- Size:25 L
- Cat. No.:EM1.05593.9025
- Supplier no.:1.05593.9025
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.219 g/cm3 (20 °C)
- pH:>11 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:119 - 135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.219 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548