
Potassium carbonate sesquihydrate, Suprapur® for trace metal analysis, Supelco®
Catalog # EM1.04926.0050
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:50 g
- Cat. No.:EM1.04926.0050
- Supplier no.:1.04926.0050
Specifications
Specification Test Results
- Solubility:1300 g/l (20 °C)
- Melting Point:891 °C (anhydrous substance)
- Molar Mass:138.21 g/mol (anhydrous)
- pH:(50 g/l, H2O, 20 °C) strongly alkaline
Specification Test Results
About this item
:
:165.23 g/mol
Melting Pt:891 °C
Density:2.29 g/cm³ (20 °C)
EINECS:209-529-3
:GHS07
Storage Temperature:Ambient
CAS Number:6381-79-9
Formula:CK₂O₃·1,5H₂O
MDL Number:MFCD00149925