
Potassium chloride 2.97-3.03 M saturated with silver chloride aqueous solution for ion chromatography, Supelco®
Catalog # EM1.04818.0100
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 ml
- Cat. No.:EM1.04818.0100
- Supplier no.:1.04818.0100
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:100 °C (1013 hPa)
- Density:1.13 g/cm3 (20 °C)
Specification Test Results
About this item
:
:74.55 g/mol
Density:1.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
UN:3082
ADR:9,III
:GHS09
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360