Order Entry
Puerto Rico
ContactUsLinkComponent
 
 
 
Hydrochloric acid 0.1000 M in 2-propanol, Titripur® DIN 51558-1, Supelco®
  EM1.00326.1000
 :  
 
 
Hydrochloric acid 0.1000 M in 2-propanol, Titripur® DIN 51558-1, Supelco®
  EM1.00326.1000
 :  1.00326.1000
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    EM1.00326.1000
  • Supplier no.:
    1.00326.1000

 

 

  • Solubility:
    (20 °C) soluble
  • Flash Point:
    12 °C (2-Propanol)
  • Explosion Limit:
    2 - 13.4 %(V) (2-Propanol)
  • Density:
    0.79 g/cm3 (20 °C)
  • pH:
    < - 0.5 (H2O, 20 °C)
  • Ignition Point:
    425 °C (2-Propanol)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

 

 

 :
 :0.79 g/cm³ (20 °C)
 :231-595-7
 :3 (8),II
 :36.46
 :2924
 :GHS02
 :GHS07
 :Ambient
 :7647-01-0
 :HCl
 :MFCD00011324