
Diboron trioxide for analysis of silicates, Supelco®
Catalog # EM1.00163.1000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:EM1.00163.1000
- Supplier no.:1.00163.1000
Specifications
Specification Test Results
- Solubility:36 g/l (25 °C)
- Melting Point:450 °C
- Molar Mass:69.61 g/mol
- Bulk Density:900 - 1100 kg/m3
- Boiling Point:1860 °C
- Density:2.46 g/cm3 (20 °C)
- pH:4 (10 g/l, H2O, 25 °C)
Specification Test Results
About this item
:
:69.62 g/mol
Merck Index:13,01327
Boiling Pt:1860 °C (1013 hPa)
Boiling Pt:1860 °C (1013 hPa)
Melting Pt:450 °C
Density:1.81 to 1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-125-8
REACH:01-2119486655-24
:GHS08
CAS Number:1303-86-2
Formula:B₂O₃
MDL Number:MFCD00011315