FIREBOY Safety Bunsen Burner, INTEGRA Biosciences
Supplier: Avantor
Certificates
About this item
The FIREBOY™ burner ensures highest application safety by providing continuous flame monitoring, overtemperature protection and user-defined, maximal burning times.
- Compatible to common gas types such as butane/propane and natural gas
- Smooth, chrome-plated metal housing is easy to clean
- UV- and solvent-resistant
If equipped with a rechargeable battery (optional for model 144000) and a gas cartridge adapter, the FIREBOY is independent of any gas distribution system and power supply, allowing the unit to be used wherever required. Depending on the model, the unit can be operated in three different operation modes – button, sensor or footswitch mode. The automatic ignition, triggered by the optical sensor or the foot switch, allows hands-free operation.
100 - 240 V AC, 50/60 Hz
Propane/Butane: 50 mbar, Natural Gas: 20 mbar (gas input pressure)
100×150 mm (3.94×5.9")
1.3 kg (2.87 lbs.)
Certifications: CE certified.
Ordering information: Units include mains adapter, 2 nozzles for propane/butane (p60) and natural gas (n80), connector for 10 mm gas tubing and footswitch (only for model 144010).