SP Bel-Art Poly-Hand Pumps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
The Poly-Hand pump threads directly to a standard drum bung or carboy.
- Polyethylene construction
- Rugged and inert
- Ergonomically designed handle for comfortable use
It is constructed entirely of rugged polyethylene and is inert to strong acids, alkalis and many solvents
The pump delivers by positive pressure on the downstroke of the piston and does not pressurize the vessel. Its ergonomic 'T' handle fits comfortably in a gloved hand. The pump will efficiently deliver against a 3.65 m (12 ft) head of water and empty a drum or carboy to within 3 mm (⅛") of the bottom.
Sizes are available to fit 114 L and 220 L (30/50 gal) plastic or steel drums and 20 L (5 gal.) carboy (16126-845 ).