ContactUsLinkComponent
TitroLine® 7000 Titrators, SI Analytics
 :  YSI
undefined
TitroLine® 7000 Titrators, SI Analytics
 :  YSI
  • Catalog No:
  • 76403-700
  • Measuring Range:
  • −3.0 - 18.0…0.002 ±1 digit
  • Display:
  • Graphics-capable TFT display
  • Keypad:
  • PC-keyboard (USB)
  • Stirrer connection:
  • TM 50, power supply directly through piston burette
  • Dosing Accuracy:
  • 0.15%, precision <0.05 - 0.07% in compliance with EN ISO 8655-3
  • Burette Resolution:
  • 2000
  • pH Accuracy:
  • −3.0 - 18.0…0.002 ±1 digit
  • mV range:
  • −2000 - 2000…0.1 ±1 digit
  • Power supply:
  • 100 - 240 V
  • Operating Temperature:
  • 10...40 °C
  • W×D×H:
  • 15.3×29.6×45 cm
  • Weight:
  • 3.5 kg