ContactUsLinkComponent
TITRONIC® 500 Titrators, SI Analytics
 :  YSI
undefined
TITRONIC® 500 Titrators, SI Analytics
 :  YSI
  • Catalog No:
  • 76403-678
  • Display:
  • 3.5" - 1/4 VGA TFT graphic color display with 320×240 pixels
  • Dosing Accuracy:
  • In compliance with EN ISO 8655 part 3, systematic error <0.15%, random error <0.05 - 0.07%
  • Power supply:
  • 100 - 240 V, 50/60 Hz
  • Operating Temperature:
  • 10...40 °C (50...104 °F)
  • W×D×H:
  • 15.3×29.6×45 cm
  • Weight:
  • 3.5 kg (7.7 lbs.) with exchangeable head