Orders LinkContactUsLinkComponent
Inoculation Loop, Globe Scientific
Inoculation Loop, Globe Scientific
Inoculation Loop, Globe Scientific
Inoculation Loop, Globe Scientific
Inoculation Loop, Globe Scientific
Loops offer the most convenient and safest way of streaking culture media
  • Polystyrene (PS)
  • Certificate of Calibration comes with each case of product
  • Gamma sterilized
  • Rigid feel for comfortable, stable, uniform streaking
  • One side is a 10 µl loop and the reverse side is a 1 µl loop
Loops are sterilized by gamma radiation.
Molded from a special medical grade polystyrene, they offer a strong, rigid feel to provide comfortable, stable, and uniform streaking.
Delivery information: Supplied with calibration certificate.
Documents
Product Family Options
  • Description
    Color
    Sterility
    Packaging
  • Inoculation Loop, 1 uL and 10 uL
    White
    Sterile
    10/Peel Pack, 50 Packs/Case
    Case of 500
    Case of 500 - $98.06
    $98.06
    Case of 500
       $98.06
     : 2860
  • Inoculation Loop, 1 uL and 10 uL
    White
    Sterile
    Individually Wrapped
    Case of 500
    Case of 500 - $253.18
    $253.18
    Case of 500
       $253.18
     : 2850
  • Inoculation Loop, 1 uL and 10 uL
    White
    Sterile
    20/Peel Pack, 25 Packs/Case
    Case of 500
    Case of 500 - $87.54
    $87.54
    Case of 500
       $87.54
     : 2810