Order Entry
Puerto Rico
ContactUsLinkComponent
Protease (from Staphylococcus aureus, Strain V8), MP Biomedicals
Protease (from Staphylococcus aureus, Strain V8), MP Biomedicals
Catalog # IC399001
Supplier:  MP Biomedicals
CAS Number:  
undefined
Protease (from Staphylococcus aureus, Strain V8), MP Biomedicals
Catalog # IC399001
Supplier:  MP Biomedicals
Supplier Number:  08399001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
    Staphylococcus aureus, Strain V8
  • Size:
    5 mg
  • Storage Conditions:
    Store at 2...8 °C, –20 °C for Extended Storage
  • Enzyme Name:
    Protease
  • pH:
    pH Optimal: 4.0 and 7.8 (Hemoglobin Substrate)
  • Cat. No.:
    IC399001
  • Supplier no.:
    399001

Specifications

About this item

Protease V.8 specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids.

  • Lyophilized powder that has been chromatographically purified
  • Specific activity: 915 units/mg (dry weight)
  • pH Optimal: 4.0 and 7.8 (hemoglobin substrate)
  • Robust activity, greater than 915 units/mg protein
  • Specific cleavage for glutamic acid in ammonium buffer
  • Selective digestion at both glutamic and aspartic acid residues in phosphate buffer at pH 7.8
  • Cleavage activity remains in the presence of many denaturing agent, such as SDS, urea and guanidine-HCl
  • Lyophilized format for longer stability and easier storage

In the presence of ammonium, the specificity is limited to glutamic sites. The enzyme specifically cleaves peptide bonds on the COOH terminal side of either aspartic or glutamic acids. It is used in a study to investigate immobilized residue-specific endoproteinases for protein sequencing. Protease V8 is used in a study to assess enzymatic cleaning of inorganic ultrafiltration membranes used for whey protein fractionation.

The enzyme was isolated, by the method of Drapeau, from the culture filtrate of Staphylococcus aureus V.8 strain. The preparation is homogenous on ultracentrifugation and disc electrophoresis.