Pokeweed mitogen, MP Biomedicals
Catalog # IC780331
Supplier: MP Biomedicals
CAS Number:
Specifications
- Size:5 mg
- Protein/Peptide Name:Pokeweed mitogen
- Cat. No.:IC780331
- Supplier no.:780331
Specifications
About this item
Form: Lyophilized powder.
Reconstitution: Reconstitute with 0.01M sodium phosphate-0.15M sodium chloride, pH 7.2-7.4.
Activity: Shows agglutination when tested with human red blood cells.