ContactUsLinkComponent
Phytohemagglutinin, MP Biomedicals
Phytohemagglutinin, MP Biomedicals
  IC791501
 :  
Phytohemagglutinin, MP Biomedicals
  IC791501
 :  08791501
 :  
  • Size:
    25 mg
  • Protein/Peptide Name:
    Phytohemagglutinin
  • Cat. No.:
    IC791501
  • Supplier no.:
    791501
Form: Lyophilized material.
Description: Crude Phaseolus vulgaris-P lectin (PHA-P) from the Red Kidney Bean. Phytohemagglutinin PHA agglutinates the erythrocytes of all human blood groups as well as those of rabbit, dog, cat, mouse, sheep, and guinea pig.