Order Entry
United States
ContactUsLinkComponent
 
ImmunO™ Ouchterlony Plates, MP Biomedicals
Catalog # IC642781
Supplier:  MP Biomedicals
ImmunO™ Ouchterlony Plates, MP Biomedicals
Catalog # IC642781
Supplier:  MP Biomedicals
Supplier Number:  08642781
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • No. of wells:
    56
  • Well volume:
    10 μL
  • Description:
    Ouchterlony Plates, Max. Sample Volume 10 μl per well
  • Environmentally Preferable:
  • Cat. No.:
    IC642781
  • Packaging:
    5 Plates
  • Supplier no.:
    642781

Specifications

About this item

The MP ouchterlony plate is designed for specific but low affinity antisera

  • Typically dilutions of 1:600 to 1:16,000 are required

Each MP immunodiffusion plate contains 8 ouchterlony patterns consisting of a center well and 6 outer wells.The high quality immunodiffusion grade agarose contains 5% PEG which greatly enhances the sensitivity of the immunodiffusion assay. The plates contain 1.2% agarose (buffer - PBS with 0.1% sodium azide and 0.01% merthiolate). The maximum sample volume is 10 ul in any of the wells.

Such antisera require properly spaced wells and olyethylene glycol in the agarose for optimal immuno -precipitation.