Order Entry
Puerto Rico
ContactUsLinkComponent
 
Agarose Immunodiffusion Grade, MP Biomedicals
 
undefined
Agarose Immunodiffusion Grade, MP Biomedicals

 

  • Catalog No:
  • IC952014
  • IC952012
  • Gel Strength:
  • 2890 g/cm2 with 1.5% gel
  • Solubility:
  • Soluble in Water
  • CAS No.:
  • 9012-36-6
  • Form:
  • Lyophilized powder