Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB), powder

Supplier:   MP Biomedicals
3,3',5,5'-Tetramethylbenzidine (TMB), powder
Supplier:  MP Biomedicals
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB), powder
3,3',5,5'-Tetramethylbenzidine (TMB), powder

2 Options Available Below

Product Details & Documents

(TMB Powder) This product is excellent for ELISA procedures.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10×100 mg
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $399.00
    $399.00
    Each (100µG)
       $399.00
     

    Supplier Number: 08980502
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $297.00
    $297.00
    Each (5g)
       $297.00
     

    Supplier Number: 08980501