Trypsin
500 pmol
Catalog # 101641-072
Small Business Enterprise
Supplier: New England Biolabs (NEB)
Supplier #: P8108S
Trypsin
Catalog # 101641-072
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
A complex mixture of peptides produced by Trypsin digestion of Bovine Serum Albumin (BSA) that was reduced and alkylated with Iodoacetamide (CAM modified)
This peptide mixture can be used to test a Matrix-Assisted Laser Desorption/Ionization Time-Of-Flight (MALDI-TOF) or Electrospray Ionization (ESI) mass spectrometer (TOF, Q-TOF or Ion Trap).
Specifications
Specifications
- Size:500 pmol
- Cat. No.:101641-072
- Enzyme Name:Trypsin