- Description:TriDye 100 bp DNA Ladder
- Size:125 gel lanes
- Environmentally Preferable:
- Cat. No.:101227-144
- Supplier no.:N3271S
TriDye™ 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing 3 dyes which serve as visual aids to monitor the progress of migration during agarose gel electrophoresis
The DNA ladder consists of proprietary plasmids are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference points.