SphI
Catalog # 101228-944
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Specifications
- Concentration:80000 units/ml
- Description:SphI
- Size:2500 units
- Enzyme Name:SphI
- Cat. No.:101228-944
- Supplier no.:R0182M
Specifications
About this item
An E.coli strain that carries the SphI gene from Streptomyces phaeochromogenes (NRRL B-3559).