Order Entry
United States
ContactUsLinkComponent
SOC Outgrowth Medium, New England Biolabs
SOC Outgrowth Medium, New England Biolabs
Catalog # 101417-886
Small Business Enterprise
CAS Number:  
undefined
SOC Outgrowth Medium, New England Biolabs
Catalog # 101417-886
Supplier Number:  B9020S
Small Business Enterprise
CAS Number:  

Specifications

  • Source:
  • Species:
  • Description:
    SOC Outgrowth Medium
  • Size:
    100 mL
  • Environmentally Preferable:
  • Cat. No.:
    101417-886
  • Supplier no.:
    B9020S

Specifications

About this item

Add SOC to freshly transformed E.coli cells and incubate for up to 1 hour before plating on selective media.