ContactUsLinkComponent
SNAP-SURFACE® 594, New England Biolabs
SNAP-SURFACE® 594, New England Biolabs
Catalog # 101447-682
Small Business Enterprise
CAS Number:  
undefined
SNAP-SURFACE® 594, New England Biolabs
Catalog # 101447-682
Supplier Number:  S9134S
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    SNAP-Surface 594
  • Size:
    50 nmol
  • Cat. No.:
    101447-682
  • Supplier no.:
    S9134S
Specifications
About this item
SNAP-Surface® 594 is a photostable fluorescent substrate that can be used to label SNAP-tag® fusion proteins in solution or on the surface of living or fixed cells
This substrate (BG-594) is based on the ATTO-TEC dye ATTO 594 and is suitable for standard Texas Red and Alexa 594 filter sets. It has an excitation maximum at 606 nm and emission maximum at 626 nm. This package includes 50 nmol of SNAP-Surface 594 substrate, sufficient to make 10 ml of a 5 µM SNAP-tag fusion protein labeling solution.