Specifications
- Description:SNAP-Cell TMR-STAR
- Size:30 nmol
- Cat. No.:101447-660
- Supplier no.:S9105S
Specifications
About this item
SNAP-Cell® TMR-Star is a red fluorescent substrate that can be used to label SNAP-tag® fusion proteins inside living cells, on cell surfaces, or in vitro
This cell-permeable substrate (CP-6-TMR) is based on the single isomer 6-carboxytetramethylrhodamine and is suitable for standard rhodamine filter sets. It has an excitation maximum at 554 nm and an emission maximum at 580 nm. This package contains 30 nmol of SNAP-Cell TMR-Star substrate, sufficient to make 10 ml of a 3 µM SNAP-tag fusion protein labeling solution.