Order Entry
Puerto Rico
ContactUsLinkComponent
Ribonucleoside vanadyl complex 200 mM
Ribonucleoside vanadyl complex 200 mM
Catalog # 101229-716
Small Business Enterprise
CAS Number:  
undefined
Ribonucleoside vanadyl complex 200 mM
Catalog # 101229-716
Supplier Number:  S1402S
Small Business Enterprise
CAS Number:  

Specifications

  • Size:
    10 ml
  • Cat. No.:
    101229-716
  • Supplier no.:
    S1402S

Specifications

About this item

 :
Storage Temperature:Freezer
 :123334-20-3