- Description:PURExpress (aa, tRNA) Kit
- Size:10 Reactions
- Cat. No.:101641-562
- Supplier no.:E6840S
A rapid method for gene expression analysis, PURExpress® is a novel cell-free transcription/translation system reconstituted from the purified components necessary for E
The relative nuclease-free and protease-free nature of the PURExpress platform preserves the integrity of DNA and RNA templates/ complexes and results in proteins that are free of modification and degradation. Transcription and translation are carried out in a one-step reaction, and require the mixing of only two tubes. With results available in a few hours, PURExpress saves valuable laboratory time and is ideal for high throughput technologies. The tRNA and amino acids are supplied separately in this kit, allowing users to run a protein synthesis reaction by adding modified amino acids and tRNA mixtures to the reaction.
coli translation.