Specifications
- Description:pSNAP-tag (T7)-2 Vector
- Size:20 µg
- Cat. No.:101641-578
- Supplier no.:N9181S
- Vector details:pSNAP
Specifications
About this item
pSNAP-tag (T7)-2 Vector is an Escherichia coli expression plasmid encoding the SNAP-tag protein
Expression is under control of the IPTG inducible T7 promoter. The pSNAP-tag (T7)-2 Vector can also be used as a control plasmid expressing the SNAP-tag protein (20 kDa). The target gene should be cloned as a fusion to the N- or C-terminus of the SNAP-tag.