E. faecalis O-Glycosidase and Neuraminidase Bundle
102500-120
Small Business Enterprise
:
- Source:E. coli
- Species:E. faecalis
- Size:2000 units
- Enzyme Name:O-Glycosidase and Neuraminidase Bundle
- Cat. No.:102500-120
- Supplier no.:E0540S
1 set of this bundle includes 2,000,000 units of O-Glycosidase and 2,000 units of Neuraminidase
O-Glycosidase, also known as Endo-α-N-Acetylgalactosaminidase, catalyzes the removal of Core 1 and Core 3 O-linked disaccharides from glycoproteins. Neuraminidase is the common name for Acetyl-neuraminyl hydrolase (Sialidase). This Neuraminidase catalyzes the hydrolysis of α2-3, α2-6, and α2-8 linked N-acetyl-neuraminic acid residues from glycoproteins and oligosaccharides.